4-Chloro-2,3-difluoropyridine ≥97%

Supplier: AOBChem USA

Warning

11512-1G 11512-25G 11512-500MG 11512-5G
76924-602EA 263.11 USD
76924-602 76924-604 76924-606 76924-608
4-Chloro-2,3-difluoropyridine ≥97%
4-Chloro-2,3-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂ClF₂N
MW: 149.526 g/mol
Storage Temperature: Ambient
CAS Number: 851178-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR