5-Chloro-2-methoxy-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-2-methoxypyridine-3-boronic acid, (5-Chloro-2-methoxypyridin-3-yl)boronic acid

11453-25G 11453-5G
76924-578EA 350.1 USD
76924-578 76924-580
5-Chloro-2-methoxy-3-pyridineboronic acid ≥97%
5-Chloro-2-methoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₃
MW: 187.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114579
CAS Number: 943153-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR