4-Cyano-2-nitrophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Cyano-2-nitrophenyl)boronic acid

11439-1G 11439-25G 11439-5G
76924-564EA 62.6 USD
76924-564 76924-566 76924-568
4-Cyano-2-nitrophenylboronic acid ≥97%
4-Cyano-2-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BN₂O₄
Storage Temperature: Ambient
MDL Number: MFCD04115646
CAS Number: 850568-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR