(5-Chloro-2-isopropoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-2-isopropoxyphenylboronic acid

11334-25G 11334-5G
76924-546EA 233.4 USD
76924-546 76924-548
(5-Chloro-2-isopropoxyphenyl)boronic acid ≥97%
(5-Chloro-2-isopropoxyphenyl)boronic acid
Formula: C₉H₁₂BClO₃
MW: 214.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06411283
CAS Number: 352534-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR