5-Chloro-2-fluoro-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

11295-1G 11295-25G 11295-500MG 11295-5G
76924-534EA 260.99 USD
76924-534 76924-536 76924-538 76924-540
5-Chloro-2-fluoro-4-methoxyphenylboronic acid ≥97%
5-Chloro-2-fluoro-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClFO₃
MW: 204.391 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04112552
CAS Number: 1072952-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR