4-Bromo-2-ethoxy-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-ethoxytolune

11232-25G 11232-5G
76924-506EA 375.56 USD
76924-506 76924-508
4-Bromo-2-ethoxy-1-methylbenzene ≥97%
4-Bromo-2-ethoxy-1-methylbenzene
Formula: C₉H₁₁BrO
MW: 215.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617247
CAS Number: 871888-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR