4-Bromo-2-isopropylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

11154-1G 11154-25G 11154-500MG 11154-5G
76924-494EA 195.21 USD
76924-494 76924-496 76924-498 76924-500
4-Bromo-2-isopropylbenzaldehyde ≥97%
4-Bromo-2-isopropylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO
Storage Temperature: Refrigerator
MDL Number: MFCD09835100
CAS Number: 1114808-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR