4-Bromo-2-fluoro-1-isopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

110885-1G 110885-25G 110885-500MG 110885-5G
76924-482EA 334.19 USD
76924-482 76924-484 76924-486 76924-488
4-Bromo-2-fluoro-1-isopropylbenzene ≥95%
4-Bromo-2-fluoro-1-isopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrF
Storage Temperature: Ambient
MDL Number: MFCD20482704
CAS Number: 112611-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR