4-Bromo-2-fluoro-1-((4-methylbenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

110323-1G 110323-25G 110323-500MG 110323-5G
76924-468EA 233.4 USD
76924-468 76924-470 76924-472 76924-474
4-Bromo-2-fluoro-1-((4-methylbenzyl)oxy)benzene ≥97%
4-Bromo-2-fluoro-1-((4-methylbenzyl)oxy)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO
MW: 295.147 g/mol
Storage Temperature: Ambient
CAS Number: 1332347-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR