4-Bromo-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10950-1G 10950-25G 10950-5G
76924-460EA 62.6 USD
76924-460 76924-462 76924-464
4-Bromo-2-fluorophenylboronic acid ≥97%
4-Bromo-2-fluorophenylboronic acid
Formula: C₆H₅BBrFO₂
Storage Temperature: Ambient
MDL Number: MFCD01318110
CAS Number: 216393-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR