5-Bromo-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

10919-25G 10919-5G
76924-450EA 346.92 USD
76924-450 76924-452
5-Bromo-2-methylbenzaldehyde ≥97%
5-Bromo-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO
Storage Temperature: Ambient
MDL Number: MFCD05662390
CAS Number: 90050-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR