5-Methoxycarbonyl-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-(Methoxycarbonyl)-2-methylphenyl)boronic acid, 5-(Methoxycarbonyl)-2-methylphenylboronic acid

10820-1G 10820-25G 10820-5G
76924-432EA 79.57 USD
76924-432 76924-434 76924-436
5-Methoxycarbonyl-2-methylphenylboronic acid ≥97%
5-Methoxycarbonyl-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MW: 193.992 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111429
CAS Number: 876189-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR