4-Bromo-5-fluorobenzene-1,2-diol ≥97%
Supplier: AOBChem USA
|
Warning
|
107738-1G
107738-25G
107738-500MG
107738-5G
76924-418EA
385.11
USD
76924-418
76924-420
76924-422
76924-424
4-Bromo-5-fluorobenzene-1,2-diol ≥97%
4-Bromo-5-fluorobenzene-1,2-diol
Precaución: for Research Use Only
Formula:
C₆H₄BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD13177846 CAS Number: 656804-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...