4-Bromo-3-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-3-fluorobenzeneboronic acid

10766-25G 10766-5G
76924-412EA 488.02 USD
76924-412 76924-414
4-Bromo-3-fluorophenylboronic acid ≥97%
4-Bromo-3-fluorophenylboronic acid
Formula: C₆H₅BBrFO₂
MW: 218.82 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093066
CAS Number: 374790-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR