5-Acetyl-2-methoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (5-Acetyl-2-methoxyphenyl)boronic acid

10701-25G 10701-5G
76924-406EA 630.18 USD
76924-406 76924-408
5-Acetyl-2-methoxyphenylboronic acid ≥95%
5-Acetyl-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MW: 193.992 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839106
CAS Number: 1215281-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR