5-Bromothiophene-2-carboxamide ≥97%

Supplier: AOBChem USA

Warning

106269-25G 106269-5G
76924-396EA 456.19 USD
76924-396 76924-398
5-Bromothiophene-2-carboxamide ≥97%
5-Bromothiophene-2-carboxamide
Formula: C₅H₄BrNOS
Storage Temperature: Ambient
MDL Number: MFCD01922020
CAS Number: 76371-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR