4-Ethoxycarbonyl-3-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

10573-1G 10573-25G 10573-500MG 10573-5G
76924-386EA 129.43 USD
76924-386 76924-388 76924-390 76924-392
4-Ethoxycarbonyl-3-methoxyphenylboronic acid ≥97%
4-Ethoxycarbonyl-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BO₅
Storage Temperature: Refrigerator
MDL Number: MFCD08274475
CAS Number: 911312-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR