4-Fluoro-(3-methoxycarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid, 4-Fluoro-3-methoxycarbonylphenylboronic acid

10554-25G 10554-5G
76924-378EA 204.76 USD
76924-378 76924-380
4-Fluoro-(3-methoxycarbonyl)phenylboronic acid ≥97%
4-Fluoro-(3-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₄
MW: 197.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06801688
CAS Number: 874219-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR