N-(4-Bromophenyl)-2,2-dimethylpropanamide ≥97%

Supplier: AOBChem USA

Warning

10212-25G 10212-5G
76924-266EA 622.75 USD
76924-266 76924-268
N-(4-Bromophenyl)-2,2-dimethylpropanamide ≥97%
N-(4-Bromophenyl)-2,2-dimethylpropanamide
Formula: C₁₁H₁₄BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD00124399
CAS Number: 24109-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR