5-Cyano-2-fluorophenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
5-Cyano-2-fluorobenzeneboronic acid
10195-25G
10195-5G
76924-260EA
219.61
USD
76924-260
76924-262
5-Cyano-2-fluorophenylboronic acid ≥97%
5-Cyano-2-fluorophenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₅BFNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD03095147 CAS Number: 468718-30-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...