5-(Benzyloxy)-2-bromobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

10173-25G 10173-5G
76924-238EA 566.53 USD
76924-238 76924-240
5-(Benzyloxy)-2-bromobenzaldehyde ≥97%
5-(Benzyloxy)-2-bromobenzaldehyde
Formula: C₁₄H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD03425931
CAS Number: 85604-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR