5-Bromo-2-fluoro-3-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-fluoropyridine-3-boronic acid pinacol ester, 5-Bromo-2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

10148-25G 10148-5G
76924-220EA 289.63 USD
76924-220 76924-222
5-Bromo-2-fluoro-3-pyridineboronic acid pinacol ester ≥97%
5-Bromo-2-fluoro-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₄BBrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD08689619
CAS Number: 1073353-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR