2-Fluoro-4-hydroxy-N-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

99950-1G 99950-25G 99950-500MG 99950-5G
76906-166EA 349.04 USD
76906-166 76906-168 76906-170 76906-172
2-Fluoro-4-hydroxy-N-methylbenzamide ≥95%
2-Fluoro-4-hydroxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16998328
CAS Number: 1243392-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR