2,6-Bis(chloromethyl)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

99889-25G 99889-5G
76906-158EA 310.85 USD
76906-158 76906-160
2,6-Bis(chloromethyl)pyridine ≥97% research grade
2,6-Bis(chloromethyl)pyridine
Formula: C₇H₇Cl₂N
Storage Temperature: Refrigerator
MDL Number: MFCD00142844
CAS Number: 3099-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR