2-Bromo-5-fluoro-4-methoxyaniline ≥97%

Supplier: AOBChem USA

Warning

97877-1G 97877-25G 97877-500MG 97877-5G
76906-144EA 141.1 USD
76906-144 76906-146 76906-148 76906-150
2-Bromo-5-fluoro-4-methoxyaniline ≥97%
2-Bromo-5-fluoro-4-methoxyaniline
Formula: C₇H₇BrFNO
MW: 220.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22200117
CAS Number: 445441-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR