2-Bromo-3,6-dimethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

95313-1G 95313-500MG 95313-5G
76906-132EA 471.04 USD
76906-132 76906-134 76906-136
2-Bromo-3,6-dimethoxybenzoic acid ≥97%
2-Bromo-3,6-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₄
MW: 261.069 g/mol
Storage Temperature: Ambient
CAS Number: 1605281-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR