2-Bromo-3,5-difluorophenol ≥95%

Supplier: AOBChem USA

Warning

95078-25G 95078-5G
76906-120EA 215.37 USD
76906-120 76906-122
2-Bromo-3,5-difluorophenol ≥95%
2-Bromo-3,5-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861127
CAS Number: 325486-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR