2-Bromo-5-(diethoxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

92930-25G 92930-5G
76906-108EA 346.92 USD
76906-108 76906-110
2-Bromo-5-(diethoxymethyl)pyridine ≥97%
2-Bromo-5-(diethoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrNO₂
MW: 260.128 g/mol
Storage Temperature: Refrigerator
CAS Number: 1350349-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR