2,6-Diisopropyliodobenzene ≥95%

Supplier: AOBChem USA

Warning

90602-1G 90602-25G 90602-5G
76906-070EA 92.3 USD
76906-070 76906-072 76906-074
2,6-Diisopropyliodobenzene ≥95%
2,6-Diisopropyliodobenzene
Formula: C₁₂H₁₇I
Storage Temperature: Ambient
MDL Number: MFCD12547790
CAS Number: 163704-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR