2-Bromo-6-chloro-4-ethylphenol ≥97%

Supplier: AOBChem USA

Warning

86617-1G 86617-500MG 86617-5G
76906-028EA 530.45 USD
76906-028 76906-030 76906-032
2-Bromo-6-chloro-4-ethylphenol ≥97%
2-Bromo-6-chloro-4-ethylphenol

Precaución: for Research Use Only

Formula: C₈H₈BrClO
Storage Temperature: Ambient
CAS Number: 57018-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR