2-Chloro-4-iodo-6-methylaniline ≥97%

Supplier: AOBChem USA

Warning

83986-25G 83986-5G
76906-018EA 346.92 USD
76906-018 76906-020
2-Chloro-4-iodo-6-methylaniline ≥97%
2-Chloro-4-iodo-6-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇ClIN
MW: 267.495 g/mol
Storage Temperature: Ambient
CAS Number: 156746-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR