2-Ethoxy-3-ethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

83446-1G 83446-25G 83446-500MG 83446-5G
76906-006EA 266.29 USD
76906-006 76906-008 76906-010 76906-012
2-Ethoxy-3-ethylbenzaldehyde ≥97%
2-Ethoxy-3-ethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.228 g/mol
Storage Temperature: Ambient
CAS Number: 2018745-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR