2-Chloro-3-fluoroisonicotinamide ≥97%

Supplier: AOBChem USA

Warning

81015-1G 81015-500MG 81015-5G
76905-976EA 665.19 USD
76905-976 76905-978 76905-980
2-Chloro-3-fluoroisonicotinamide ≥97%
2-Chloro-3-fluoroisonicotinamide

Precaución: for Research Use Only

Formula: C₆H₄ClFN₂O
MW: 174.56 g/mol
Storage Temperature: Ambient
CAS Number: 1378654-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR