2-Bromo-3-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

80581-25G 80581-5G
76905-950EA 590.93 USD
76905-950 76905-952
2-Bromo-3-methoxyphenol ≥97%
2-Bromo-3-methoxyphenol
Formula: C₇H₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD09056758
CAS Number: 135999-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR