2-Bromo-3-fluoro-4-iodo-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

79914-25G 79914-5G
76905-938EA 392.54 USD
76905-938 76905-940
2-Bromo-3-fluoro-4-iodo-1-methylbenzene ≥97%
2-Bromo-3-fluoro-4-iodo-1-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
Storage Temperature: Ambient
CAS Number: 1804910-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR