1-Bromo-2,6-dimethoxy-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

78626-1G 78626-500MG 78626-5G
76905-924EA 476.35 USD
76905-924 76905-926 76905-928
1-Bromo-2,6-dimethoxy-4-iodobenzene ≥97%
1-Bromo-2,6-dimethoxy-4-iodobenzene
Formula: C₈H₈BrIO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28669899
CAS Number: 1318133-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR