2-Chloro-5-fluoro-3-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

77712-1G 77712-500MG 77712-5G 77712-25G
76905-910EA 857.21 USD
76905-910 76905-912 76905-914 77660-334
2-Chloro-5-fluoro-3-isopropoxybenzaldehyde ≥97%
2-Chloro-5-fluoro-3-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₂
MW: 216.637 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34767206
CAS Number: 2918833-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR