2,6-Bis(benzyloxy)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

77112-25G 77112-5G
76905-900EA 559.1 USD
76905-900 76905-902
2,6-Bis(benzyloxy)pyridine ≥97% research grade
2,6-Bis(benzyloxy)pyridine
Formula: C₁₉H₁₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD05863260
CAS Number: 16727-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR