2-Fluoro-3,4-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

75136-1G 75136-25G 75136-500MG 75136-5G
76905-862EA 211.12 USD
76905-862 76905-864 76905-866 76905-868
2-Fluoro-3,4-dimethylbenzoic acid ≥97%
2-Fluoro-3,4-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂
Storage Temperature: Refrigerator
CAS Number: 1427391-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR