2-Iodo-3-methoxybenzotrifluoride ≥97%
Supplier: AOBChem USA
|
Warning
|
74908-1G
74908-500MG
74908-5G
74908-25G
76905-850EA
503.93
USD
76905-850
76905-852
76905-854
77656-866
2-Iodo-3-methoxybenzotrifluoride ≥97%
2-Iodo-3-methoxybenzotrifluoride
Formula:
C₈H₆F₃IO Storage Temperature: Refrigerator |
MDL Number:
MFCD18398113 CAS Number: 1261501-08-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...