2-Bromo-3-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

74499-1G 74499-25G 74499-500MG 74499-5G
76905-836EA 245.07 USD
76905-836 76905-838 76905-840 76905-842
2-Bromo-3-phenylpyridine ≥97%
2-Bromo-3-phenylpyridine
Formula: C₁₁H₈BrN
Storage Temperature: Ambient
MDL Number: MFCD04114131
CAS Number: 32864-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR