2,6-Difluoro-4-isobutoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

74092-1G 74092-25G 74092-500MG 74092-5G
76905-824EA 245.07 USD
76905-824 76905-826 76905-828 76905-830
2,6-Difluoro-4-isobutoxybenzaldehyde ≥97%
2,6-Difluoro-4-isobutoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₂
MW: 214.209 g/mol
Storage Temperature: Refrigerator
CAS Number: 2918866-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR