2-Isobutoxy-3,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

73544-1G 73544-500MG
76905-810EA 490.14 USD
76905-810 76905-812
2-Isobutoxy-3,5-dimethylbenzoic acid ≥97%
2-Isobutoxy-3,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
Storage Temperature: Ambient
CAS Number: 1369414-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR