2-Bromo-6-chloro-4-fluoro-3-methylaniline ≥97%

Supplier: AOBChem USA

Warning

72941-1G 72941-25G 72941-500MG 72941-5G
76905-794EA 228.1 USD
76905-794 76905-796 76905-798 76905-800
2-Bromo-6-chloro-4-fluoro-3-methylaniline ≥97%
2-Bromo-6-chloro-4-fluoro-3-methylaniline

Precaución: for Research Use Only

Formula: C₇H₆BrClFN
MW: 238.485 g/mol
Storage Temperature: Ambient
CAS Number: 1365988-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR