2-Bromo-3-chloro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

72293-1G 72293-25G 72293-500MG 72293-5G
76905-778EA 203.7 USD
76905-778 76905-780 76905-782 76905-784
2-Bromo-3-chloro-4-iodopyridine ≥97%
2-Bromo-3-chloro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClIN
MW: 318.338 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28734063
CAS Number: 1805015-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR