2-Bromo-3,4-difluoroanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-3,4-difluoro-1-methoxybenzene

71373-25G 71373-5G
76905-770EA 481.65 USD
76905-770 76905-772
2-Bromo-3,4-difluoroanisole ≥97%
2-Bromo-3,4-difluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
MW: 223.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261345
CAS Number: 935285-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR