2-Methyl-3-phenylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

69067-1G 69067-25G 69067-5G
76905-744EA 99.73 USD
76905-744 76905-746 76905-748
2-Methyl-3-phenylbenzoic acid ≥95%
2-Methyl-3-phenylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11554177
CAS Number: 115363-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR