2-Bromo-4-chloro-1-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

68411-25G 68411-5G
76905-736EA 524.09 USD
76905-736 76905-738
2-Bromo-4-chloro-1-(trifluoromethoxy)benzene ≥97%
2-Bromo-4-chloro-1-(trifluoromethoxy)benzene
Formula: C₇H₃BrClF₃O
MW: 275.45 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11847191
CAS Number: 1260810-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR