2-Bromo-4-chloro-3-fluoro-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

66748-1G 66748-500MG 66748-5G
76905-724EA 753.24 USD
76905-724 76905-726 76905-728
2-Bromo-4-chloro-3-fluoro-1-methoxybenzene ≥97%
2-Bromo-4-chloro-3-fluoro-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Ambient
CAS Number: 398456-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR