2-Chloro-4-fluoro-5-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

65540-1G 65540-25G 65540-500MG 65540-5G
76905-708EA 188.85 USD
76905-708 76905-710 76905-712 76905-714
2-Chloro-4-fluoro-5-methylbenzoic acid ≥95%
2-Chloro-4-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20658460
CAS Number: 208165-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR